• USA Home
  • 91930 - 2′,4′,6′-Trihydroxyacetophenone monohydrate

EMAIL THIS PAGE TO A FRIEND
91930 Sigma-Aldrich

2′,4′,6′-Trihydroxyacetophenone monohydrate

purum, ≥99.0% (HPLC)

Synonym: 2-Acetylphloroglucinol, THAP

  • CAS Number 480-66-0

  • Linear Formula (HO)3C6H2COCH3·H2O

  • Molecular Weight 186.16

  •  Beilstein Registry Number 1911197

  •  EC Number 207-556-5

  •  MDL number MFCD00149091

  •  PubChem Substance ID 24889585

Purchase

Properties

Related Categories Building Blocks, C7 to C8, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   GDSIBPPJKSBCMF-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (HPLC)
mp   218-222 °C
  219-221 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

T64602 2′,4′,6′-Trihydroxyacetophenone monohydrate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?