• USA Home
  • 92077 - 2,4,6-Triisopropylbenzenesulfonyl chloride

EMAIL THIS PAGE TO A FRIEND
92077 Sigma-Aldrich

2,4,6-Triisopropylbenzenesulfonyl chloride

purum, ≥97.0% (AT)

  • CAS Number 6553-96-4

  • Linear Formula [(CH3)2CH]3C6H2SO2Cl

  • Molecular Weight 302.86

  •  Beilstein Registry Number 1218575

  •  EC Number 229-479-6

  •  MDL number MFCD00007433

  •  PubChem Substance ID 24889600

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfonyl Halides, Sulfur Compounds More...
InChI Key   JAPYIBBSTJFDAK-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (AT)
mp   92-94 °C(lit.)
  95-97 °C
solubility   ethanol: soluble50 mg/mL, clear

Description

Other Notes

Starting material for the preparation of 2,4,6-triisopropylbenzenesulfonylhydrazide (TPSH); a convenient source of di-imine

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?