• USA Home
  • 92080 - 1-(2,4,6-Triisopropylbenzenesulfonyl)-4-nitroimidazole

EMAIL THIS PAGE TO A FRIEND
92080 Sigma-Aldrich

1-(2,4,6-Triisopropylbenzenesulfonyl)-4-nitroimidazole

purum, ≥98.0% (HPLC)

Synonym: TPSNI

  • CAS Number 63734-76-9

  • Empirical Formula (Hill Notation) C18H25N3O4S

  • Molecular Weight 379.47

  •  Beilstein Registry Number 583013

  •  MDL number MFCD00043165

  •  PubChem Substance ID 329769864

Purchase

Properties

Related Categories Chemical Biology, Chemical Synthesis, Coupling Reagents, Nucleoside Chemistry
InChI Key   ZYQZZMOLYGORFJ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
mp   157-159 °C (sealed tube)
storage temp.   −20°C

Description

Other Notes

Condensing reagent for oligonucleotide synthesis

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?