• USA Home
  • 92090 - Chlorotriisopropylsilane

EMAIL THIS PAGE TO A FRIEND
92090 Sigma-Aldrich

Chlorotriisopropylsilane

purum, ≥95.0% (GC)

Synonym: TIPSCl, Triisopropylchlorosilane, Triisopropylsilyl chloride

  • CAS Number 13154-24-0

  • Linear Formula [(CH3)2CH]3SiCl

  • Molecular Weight 192.80

  •  Beilstein Registry Number 1737446

  •  MDL number MFCD00009656

  •  PubChem Substance ID 329769865

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Chemical Synthesis, Derivatization Reagents, Derivatization Reagents GC,
vapor pressure   1.16 mmHg ( 20 °C)
InChI Key   KQIADDMXRMTWHZ-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.452(lit.)
bp   198 °C/739 mmHg(lit.)
density   0.901 g/mL at 25 °C(lit.)

Description

Other Notes

Selective silylating agent, useful in nucleotide synthesis (the TIPS group is more stable than the tert-butyldimethylsilyl group against hydrolysis)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2987 8 / PGII
WGK Germany 
3
Flash Point(F) 
147.2 °F
Flash Point(C) 
64 °C
Protocols & Articles

Articles

Silylating Reagents for GC-MS Compatible Derivatives of NSAIDs

Human and veterinary pharmaceuticals are a group of “emerging” contaminants,1, 2 some of which are produced in increasingly large volumes every year. The amounts produced are reaching quantities simi...
Luis-Alberto Martin
AnalytiX Volume 10 Article 2
Keywords: Anti-inflammatory agents, Derivatizations, Environmental, Gas chromatography, Gas chromatography mass spectrometry, Mass spectrometry, Metabolites, Pesticides, Pharmaceutical, Silylations, Veterinary

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

241725 Triisopropylsilyl chloride, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?