• USA Home
  • 92128 - 2,3,4-Trimethoxybenzaldehyde

EMAIL THIS PAGE TO A FRIEND
92128 Sigma-Aldrich

2,3,4-Trimethoxybenzaldehyde

purum, ≥98.0% (HPLC)

  • CAS Number 2103-57-3

  • Linear Formula (CH3O)3C6H2CHO

  • Molecular Weight 196.20

  •  Beilstein Registry Number 981091

  •  EC Number 218-271-0

  •  MDL number MFCD00003310

Purchase

Properties

Related Categories Aldehydes, Building Blocks, C10-C12, Carbonyl Compounds, Chemical Synthesis,
InChI Key   UCTUXUGXIFRVGX-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
refractive index   n20/D 1.5547(lit.)
bp   168-170 °C/12 mmHg(lit.)
mp   38-40 °C(lit.)
  38-41 °C
solubility   methanol: soluble0.1 g/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?