• USA Home
  • 92206 - 7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester

EMAIL THIS PAGE TO A FRIEND
92206 Sigma-Aldrich

7-Methoxy-4-coumarinylacetic acid N-succinimidyl ester

for fluorescence, ≥90% (coupling rate)

Synonym: 7-Methoxy-4-coumarinacetic acid N-succinimidyl ester, N-Succinimidyl 7-methoxy-4-coumarinylacetate

  • Empirical Formula (Hill Notation) C16H13NO7

  • Molecular Weight 331.28

  •  MDL number MFCD01863413

  •  PubChem Substance ID 329769875

Purchase

Properties

Related Categories Biochemicals and Reagents, Fluorescent Labels, Fluorescent Probes, Labels, Particles and Stains, Other Fluorescent Labels
grade   for fluorescence
InChI Key   XYBYCKCNAJSBQE-UHFFFAOYSA-N
assay   ≥90% (coupling rate)
fluorescence   λex 324 nm; λem 394 nm in 0.1 M phosphate pH 7.0
storage temp.   −20°C

Description

Other Notes

Fluorescent marker for peptides

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?