• USA Home
  • 92290 - 2,4,6-Trimethylaniline

EMAIL THIS PAGE TO A FRIEND
92290 Sigma-Aldrich

2,4,6-Trimethylaniline

purum, ≥98.0% (GC)

Synonym: Mesidine

  • CAS Number 88-05-1

  • Linear Formula (CH3)3C6H2NH2

  • Molecular Weight 135.21

  •  Beilstein Registry Number 636559

  •  EC Number 201-794-3

  •  MDL number MFCD00007740

  •  PubChem Substance ID 329769883

Purchase

Properties

InChI Key   KWVPRPSXBZNOHS-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
form   liquid (clear)
color   clear brown
refractive index   n20/D 1.551(lit.)
  n20/D 1.551
bp   108-110 °C/11 mmHg(lit.)
  233 °C(lit.)
transition temp   solidification point −5-−2 °C
density   0.963 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2810 6.1 / PGII
WGK Germany 
3
RTECS 
BZ0700000
Flash Point(F) 
204.8 °F
Flash Point(C) 
96 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

132179 2,4,6-Trimethylaniline, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?