• USA Home
  • 92515 - 2,2,4-Trimethyl-2H-imidazole 1-oxide

EMAIL THIS PAGE TO A FRIEND
92515 Sigma-Aldrich

2,2,4-Trimethyl-2H-imidazole 1-oxide

for ESR-spectroscopy, ≥98.0%

Synonym: TMIO

  • CAS Number 136440-22-7

  • Empirical Formula (Hill Notation) C6H10N2O

  • Molecular Weight 126.16

  •  Beilstein Registry Number 5424342

  •  MDL number MFCD00168351

  •  PubChem Substance ID 329770130

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Electron Spin Resonance (ESR/EPR) Spectroscopy, Spectroscopy, Spin Trapping Reagents More...
grade   for ESR-spectroscopy
InChI Key   CNIGTNPXWSLRPH-UHFFFAOYSA-N
assay   ≥98.0% (GC)
  ≥98.0%
mp   32-36 °C(lit.)
storage temp.   −20°C

Description

Application

Nitrone spin trapping agent of O-,C- and S-centered radicals.

Other Notes

Reagent employed for the spin trapping of O-,C- and S-centered radicals

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?