• USA Home
  • 92550 - 1,3,3-Trimethyl-2-methyleneindoline

EMAIL THIS PAGE TO A FRIEND
92550 Sigma-Aldrich

1,3,3-Trimethyl-2-methyleneindoline

liquid, for paper chromatography, ≥96.0%

Synonym: 2-Methylene-1,3,3-trimethylindoline, Fischer base, NSC 66176

  • CAS Number 118-12-7

  • Empirical Formula (Hill Notation) C12H15N

  • Molecular Weight 173.25

  •  Beilstein Registry Number 131162

  •  EC Number 204-235-1

  •  MDL number MFCD00005813

  •  PubChem Substance ID 329770137

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents TLC, Fluorescent agents (general),
grade   for paper chromatography
InChI Key   ZTUKGBOUHWYFGC-UHFFFAOYSA-N
assay   ≥96.0% (GC)
  ≥96.0%
form   liquid
color   red
refractive index   n20/D 1.577(lit.)
  n20/D 1.577
bp   248 °C(lit.)
density   0.979 g/mL at 25 °C(lit.)

Description

Other Notes

Lit.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
NM1926500
Flash Point(F) 
145.4 °F
Flash Point(C) 
63 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?