• USA Home
  • 92605 - Trimethyloxonium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
92605 Sigma-Aldrich

Trimethyloxonium tetrafluoroborate

purum, ≥98.0% (T)

  • CAS Number 420-37-1

  • Linear Formula (CH3)3O(BF4)

  • Molecular Weight 147.91

  •  Beilstein Registry Number 3597303

  •  EC Number 206-994-4

  •  MDL number MFCD00011798

  •  PubChem Substance ID 24889679

Purchase

Properties

Related Categories Alkyl Transfer, C-X Bond Formation (Non-Halogen), Chemical Synthesis, Synthetic Reagents
InChI Key   CZVZBKHWOFJNCR-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (T)
storage temp.   2-8°C

Description

Application

A Meerwein salt, strong electrophilic methylating agent

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Reacts violently with water.
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

281077 Trimethyloxonium tetrafluoroborate, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?