• USA Home
  • 92639 - Dextran cross-linked G-25

EMAIL THIS PAGE TO A FRIEND
92639 Sigma-Aldrich

Dextran cross-linked G-25

20-80 μm particle size

Synonym: Sephadex® alternative

Purchase

Description

Legal Information

Sephadex is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?