• USA Home
  • 92739 - 2-(Trimethylsiloxy)furan

EMAIL THIS PAGE TO A FRIEND
92739 Sigma-Aldrich

2-(Trimethylsiloxy)furan

purum, ≥97.0% (GC)

  • CAS Number 61550-02-5

  • Empirical Formula (Hill Notation) C7H12O2Si

  • Molecular Weight 156.25

  •  Beilstein Registry Number 1423066

  •  MDL number MFCD00011729

  •  PubChem Substance ID 329770162

Purchase

Properties

Related Categories Building Blocks, C4 to C7, Chemical Synthesis, Furans, Heterocyclic Building Blocks More...
InChI Key   ILBCDLOQMRDXLN-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
contains   ~0.05% 2,6-di-tert-butyl-4-methylphenol as stabilizer
refractive index   n20/D 1.436(lit.)
  n20/D 1.436
bp   34-35 °C/9 mmHg(lit.)
density   0.929 g/mL at 25 °C(lit.)
shipped in   wet ice
storage temp.   −20°C

Description

Caution

may discolor to greenish-yellow on storage

Other Notes

Reactive furan; Lewis acid mediated reaction with aldehydes and acetals to yield 4-substituted 2-buten-1,4-olides ; Reaction with imines ; Cycloaddition reactions giving fused γ-lactones

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
3
Flash Point(F) 
64.4 °F
Flash Point(C) 
18 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

308064 2-(Trimethylsiloxy)furan, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?