• USA Home
  • 92756 - Ethyl trimethylsilylacetate

EMAIL THIS PAGE TO A FRIEND
92756 Sigma-Aldrich

Ethyl trimethylsilylacetate

purum, ≥95.0% (GC)

Synonym: ETSA

  • CAS Number 4071-88-9

  • Linear Formula (CH3)3SiCH2CO2C2H5

  • Molecular Weight 160.29

  •  Beilstein Registry Number 1755902

  •  EC Number 223-783-2

  •  MDL number MFCD00009172

  •  PubChem Substance ID 24889715

Purchase

Properties

Related Categories Building Blocks, C6 to C7, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   QQFBQBDINHJDMN-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
refractive index   n20/D 1.415(lit.)
  n20/D 1.415
bp   156-159 °C(lit.)
density   0.876 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent for the synthesis of α,β-unsaturated esters by Peterson olefination ; F-catalyzed 1,4-addition to enones ; Reagent for silylations under nearly neutral conditions

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
118.4 °F
Flash Point(C) 
48 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

209120 Ethyl trimethylsilylacetate, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?