• USA Home
  • 92757 - (Trimethylsilyl)acetic acid

EMAIL THIS PAGE TO A FRIEND
92757 Sigma-Aldrich

(Trimethylsilyl)acetic acid

purum, ≥99.0% (T)

Purchase

Properties

Related Categories Building Blocks, C1 to C5, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   JDMMZVAKMAONFU-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (T)
mp   39-42 °C(lit.)
  39-42 °C

Description

Other Notes

For the synthesis of α-trimethylsilylcarboxylic acids, α,β-unsaturated acids and α-trimethylsilyl-γ-butyrolactones via the dianion; For the conversion of carbonyl compounds to α,β-unsaturated thiol esters; Preparation of TMS-ketene

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AK0843500
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?