• USA Home
  • 92781 - Tributyl(trimethylsilyl)stannane

EMAIL THIS PAGE TO A FRIEND
92781 Sigma-Aldrich

Tributyl(trimethylsilyl)stannane

purum, ≥97.0% (GC)

Synonym: (Tributylstannyl)trimethylsilane, (Trimethylsilyl)tributylstannane, (Trimethylsilyl)tributyltin

  • CAS Number 17955-46-3

  • Linear Formula [CH3(CH2)3]3SnSi(CH3)3

  • Molecular Weight 363.24

  •  Beilstein Registry Number 4127163

  •  MDL number MFCD00054903

  •  PubChem Substance ID 329770167

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins
InChI Key   MZUUBYSIDDKUKE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.488(lit.)
  n20/D 1.488
bp   102-103 °C/0.5 mmHg(lit.)
density   1.04 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent undergoing cis-addition to terminal acetylenes with tin at the internal position; Generation of tributylstannyl anions

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2788 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?