• USA Home
  • 92785 - 2-(Trimethylsilyl)thiazole

EMAIL THIS PAGE TO A FRIEND
92785 Sigma-Aldrich

2-(Trimethylsilyl)thiazole

purum, ≥97.0% (GC)

Synonym: 2-Thiazolyltrimethylsilane, 2-TST

  • CAS Number 79265-30-8

  • Empirical Formula (Hill Notation) C6H11NSSi

  • Molecular Weight 157.31

  •  Beilstein Registry Number 3538017

  •  MDL number MFCD00066274

  •  PubChem Substance ID 329770169

Purchase

Properties

Related Categories Building Blocks, C3 to C7, Chemical Synthesis, Heterocyclic Building Blocks, Thiazoles More...
InChI Key   VJCHUDDPWPQOLH-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.497(lit.)
  n20/D 1.498
bp   56-57 °C/10 mmHg(lit.)
density   0.985 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Starting material for the synthesis of 2-acylthiazoles; Formyl anion equivalent in the diastereoselective addition to aldehydes

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1993 / PGIII
WGK Germany 
3
Flash Point(F) 
143.6 °F
Flash Point(C) 
62 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?