EMAIL THIS PAGE TO A FRIEND
92830 Sigma-Aldrich

Trioctylamine

purum, ≥98.0% (GC)

Synonym: TOA

  • CAS Number 1116-76-3

  • Linear Formula [CH3(CH2)7]3N

  • Molecular Weight 353.67

  •  Beilstein Registry Number 1210618

  •  EC Number 214-242-1

  •  MDL number MFCD00009560

  •  PubChem Substance ID 24889736

Purchase

Properties

InChI Key   XTAZYLNFDRKIHJ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.449(lit.)
  n20/D 1.449
bp   164-168 °C/0.7 mmHg(lit.)
  365-367 °C(lit.)
density   0.809 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
RG8225000
Flash Point(F) 
325.4 °F
Flash Point(C) 
163 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?