EMAIL THIS PAGE TO A FRIEND
92840 Sigma-Aldrich

Trioctylphosphine

technical, ~90% (GC)

Synonym: P(Oct)3

  • CAS Number 4731-53-7

  • Linear Formula [CH3(CH2)7]3P

  • Molecular Weight 370.64

  •  Beilstein Registry Number 1776995

  •  EC Number 225-234-2

  •  MDL number MFCD00015298

  •  PubChem Substance ID 24889737

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
InChI Key   RMZAYIKUYWXQPB-UHFFFAOYSA-N
grade   technical
assay   ~90% (GC)
refractive index   n20/D 1.468(lit.)
  n20/D 1.468
bp   284-291 °C/50 mmHg(lit.)
density   0.831 g/mL at 25 °C(lit.)

Description

Caution

may become slightly turbid on storage

Other Notes

This product has been replaced by 117854-ALDRICH | P(Oct)3 - Trioctylphosphine, technical grade, 90%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1760 8 / PGII
WGK Germany 
2
RTECS 
SZ3450000
Flash Point(F) 
296.6 °F
Flash Point(C) 
147 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

117854 Trioctylphosphine, technical grade, 90%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?