EMAIL THIS PAGE TO A FRIEND
92862 Sigma-Aldrich

Glyceryl trioleate

technical, ≥60% (GC)

Synonym: (9Z)9-Octadecenoic acid 1,2,3-propanetriyl ester, 1,2,3-Tri(cis-9-octadecenoyl)glycerol, Glycerol trioleate, Glycerol triolein, Oleic acid triglyceride, Oleic triglyceride, TG(18:1(9Z)/18:1(9Z)/18:1(9Z)), Triolein

  • CAS Number 122-32-7

  • Linear Formula (C17H33COOCH2)2CHOCOC17H33

  • Molecular Weight 885.43

  •  Beilstein Registry Number 1718692

  •  EC Number 204-534-7

  •  MDL number MFCD00137563

Purchase

Properties

Related Categories Biochemicals and Reagents, Building Blocks, C12 to C63, Carbonyl Compounds, Chemical Synthesis,
InChI Key   PHYFQTYBJUILEZ-IUPFWZBJSA-N
grade   technical
assay   ≥60% (GC)
refractive index   n20/D 1.470
bp   235-240 °C/18 mmHg(lit.)
density   0.91 g/mL(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
RG1936500
Flash Point(F) 
626 °F
Flash Point(C) 
330 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

T7752 Glyceryl trioleate, ~65%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?