EMAIL THIS PAGE TO A FRIEND
93060 Sigma-Aldrich

Triphenylmethanol

purum, ≥98.0% (HPLC)

Synonym: Triphenylcarbinol, Trityl alcohol

  • CAS Number 76-84-6

  • Linear Formula (C6H5)3COH

  • Molecular Weight 260.33

  •  Beilstein Registry Number 1460837

  •  EC Number 200-988-5

  •  MDL number MFCD00004445

  •  PubChem Substance ID 24889781

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   LZTRCELOJRDYMQ-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (HPLC)
bp   360 °C(lit.)
mp   160-163 °C(lit.)
  161-165 °C
solubility   dioxane: soluble0.1 g/mL, clear

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?