• USA Home
  • 93070 - Triphenylmethanethiol

EMAIL THIS PAGE TO A FRIEND
93070 Sigma-Aldrich

Triphenylmethanethiol

purum, ≥96.0% (HPLC)

Synonym: Triphenylmethyl mercaptan, Trityl mercaptan

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
InChI Key   JQZIKLPHXXBMCA-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (HPLC)
mp   104-106 °C(lit.)
  104-109 °C

Description

Other Notes

Reagent for the mild introduction of the mercapto functionality

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
UN 3335
WGK Germany 
3
RTECS 
PB5335000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?