• USA Home
  • 93098 - Triphenylphosphine hydrobromide

EMAIL THIS PAGE TO A FRIEND
93098 Sigma-Aldrich

Triphenylphosphine hydrobromide

purum, ≥96.0% (T)

Synonym: Ph3P · HBr, Triphenylphosphonium bromide

  • CAS Number 6399-81-1

  • Linear Formula (C6H5)3P · HBr

  • Molecular Weight 343.20

  •  Beilstein Registry Number 3633387

  •  EC Number 229-012-6

  •  MDL number MFCD00035107

  •  PubChem Substance ID 24889793

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
InChI Key   CMSYDJVRTHCWFP-UHFFFAOYSA-N
grade   purum
assay   ≥96.0% (T)
mp   196 °C (dec.)(lit.)

Description

Other Notes

Mild reagent for the direct preparation of allylic phosphonium salts from allylic alcohols; Acidic catalyst for the cleavage of acetals, ethers and esters; formation of THP-ethers of tert-alcohols; Convenient and quantitative source of gaseous hydrogen bromide

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?