• USA Home
  • 93190 - N-[5-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide

EMAIL THIS PAGE TO A FRIEND
93190 Sigma-Aldrich

N-[5-Amino-6-(2,3-dichlorophenyl)-1,2,4-triazin-3-yl]-2,3-dichlorobenzamide

pharmaceutical impurity standard

Synonym: Lamotrigine Related Compound D (USP), Lamotrigine impurity F (PhEur)

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Anticonvulsants / Antiepileptics Standards, Lamotrigine Impurities, Pharmaceutical Impurities,
grade   analytical standard
InChI Key   RDUGDEWOUWFKPL-UHFFFAOYSA-N
assay   ≥90.0% (TLC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
impurities   ≤10% HPLC impurities
  ≤5% solvents
  ≤5% water
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?