• USA Home
  • 93220 - β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
93220 Sigma-Aldrich

β-Nicotinamide adenine dinucleotide 2′-phosphate reduced tetrasodium salt hydrate

≥90.0% (HPLC)

Synonym: β-NADPH, 2′-NADPH hydrate, Coenzyme II reduced tetrasodium salt, Dihydronicotinamide adenine dinucleotide phosphate tetrasodium salt, NADPH Na4, TPNH2 Na4, Triphosphopyridine nucleotide reduced tetrasodium salt

Purchase

Properties

Related Categories Biochemicals and Reagents, Bulk vials, NAD(P)(H) and Analogs, NADPH, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   WYWWVJHQDVCHKF-MPUNMZHWSA-J
assay   ≥90.0% (HPLC)
impurities   ≤12% water
absorbance ratio   A260/340 nm ≤2.45
storage temp.   −20°C

Description

Other Notes

Coenzymes and Cofactors, vol. 2: Pyridine Nucleotide Coenzymes; Studies on steroid 5α-reductase

Packaged based on solid weight.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Biochem/physiol Actions

NADPH is an electron donor and a cofactor for many redox enzymes including nitric oxide synthetase.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?