• USA Home
  • 93284 - Tris(2-carboxyethyl)phosphine hydrochloride

EMAIL THIS PAGE TO A FRIEND
93284 Sigma-Aldrich

Tris(2-carboxyethyl)phosphine hydrochloride

purum, ≥98.0% (C)

Synonym: TCEP

  • CAS Number 51805-45-9

  • Empirical Formula (Hill Notation) C9H15O6P · HCl

  • Molecular Weight 286.65

  •  Beilstein Registry Number 3724376

  •  MDL number MFCD00145469

  •  PubChem Substance ID 329770218

Purchase

Properties

Related Categories Chemical Synthesis, Other Synthetic Reagents for Reduction, Reduction, Synthetic Reagents
InChI Key   PBVAJRFEEOIAGW-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (C)
impurities   ≤0.5% water
solubility   H2O: soluble
storage temp.   2-8°C

Description

Application

Water soluble reagent, used for selective reduction of disulfides. More stable than DTT and useful in mass spectrometry applications.

Other Notes

This product has been replaced by C4706-ALDRICH | Tris(2-carboxyethyl)phosphine hydrochloride powder, ≥98%

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

75259 Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)

replaced by

Product #

Description

Add to Cart

C4706 Tris(2-carboxyethyl)phosphine hydrochloride, powder, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?