• USA Home
  • 93311 - TRIS glycine SDS buffer solution

EMAIL THIS PAGE TO A FRIEND
93311 Sigma-Aldrich

TRIS glycine SDS buffer solution

BioUltra, for electrophoresis, 10× concentrate

Purchase

Properties

Related Categories Alphabetical, BioUltra Buffers, Biochemicals and Reagents, Biological Buffers, Buffer Solutions More...
grade   for electrophoresis
product line   BioUltra
quality   10× concentrate
impurities   insoluble matter, passes filter test
cation traces   Al: ≤1 mg/kg
  As: ≤0.1 mg/kg
  Ba: ≤1 mg/kg
  Bi: ≤1 mg/kg
  Ca: ≤5 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤1 mg/kg
  K: ≤20 mg/kg
  Li: ≤1 mg/kg
  Mg: ≤1 mg/kg
  Mn: ≤1 mg/kg
  Mo: ≤1 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Sr: ≤1 mg/kg
  Zn: ≤1 mg/kg
λ   10 % in H2O
UV absorption   λ: 260 nm Amax: 0.07
  λ: 280 nm Amax: 0.06

Description

Physical form

0.25 M TRIS; 1.92 M glycine; 0.1% SDS; pH 8.6±0.1

Price and Availability


Interactive Buffer Table
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?