• USA Home
  • 93323 - Europium(III) tris[3-(heptafluoropropylhydroxymethylene)-l-camphorate]

EMAIL THIS PAGE TO A FRIEND
93323 Sigma-Aldrich

Europium(III) tris[3-(heptafluoropropylhydroxymethylene)-l-camphorate]

puriss. p.a., for NMR spectroscopy

Synonym: Tris[3-(heptafluoropropylhydroxymethylene)-l-camphorato]europium(III)

  • Empirical Formula (Hill Notation) C42H42EuF21O6

  • Molecular Weight 1193.71

  •  MDL number MFCD00080909

  •  PubChem Substance ID 24889837

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Resolution Reagents, Chiral Shift Reagents
grade   for NMR spectroscopy
InChI Key   VGLKHVQPWGFXEG-CHLNTJHBSA-K
grade   puriss. p.a.
mp   152-157 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?