• USA Home
  • 93332 - 2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine

EMAIL THIS PAGE TO A FRIEND
93332 Sigma-Aldrich

2,4,6-Tris(heptafluoropropyl)-1,3,5-triazine

analytical standard, for mass spectrometry

  • CAS Number 915-76-4

  • Empirical Formula (Hill Notation) C12F21N3

  • Molecular Weight 585.11

  •  Beilstein Registry Number 381010

  •  EC Number 213-023-8

  •  MDL number MFCD00042438

  •  PubChem Substance ID 24889842

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, MS Markers, MS Standards,
grade   analytical standard, for mass spectrometry
InChI Key   KXQUYHRRCVECPV-UHFFFAOYSA-N
assay   ≥98.0% (GC)
form   neat
application(s)   mass spectroscopy (MS): suitable
refractive index   n20/D 1.311(lit.)
bp   164-165 °C(lit.)
mp   ~25 °C
density   1.716 g/mL at 25 °C(lit.)
format   neat
storage temp.   2-8°C

Description

Other Notes

Chemical mass marker for negative ion chemical ionization mass spectrometry

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?