• USA Home
  • 93338 - Tris(dimethylamino)phosphine

EMAIL THIS PAGE TO A FRIEND
93338 Sigma-Aldrich

Tris(dimethylamino)phosphine

purum, ≥97.0% (GC)

Synonym: HMPT, Hexamethylphosphorous triamide

  • CAS Number 1608-26-0

  • Linear Formula P[N(CH3)2]3

  • Molecular Weight 163.20

  •  Beilstein Registry Number 906778

  •  MDL number MFCD00008301

  •  PubChem Substance ID 329770227

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Phosphine Compounds, Phosphine Ligands
InChI Key   XVDBWWRIXBMVJV-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
impurities   ≤0.1% hexamethylphosphoric acid triamide
refractive index   n20/D 1.463(lit.)
  n20/D 1.466
bp   48-50 °C/12 mmHg(lit.)
density   0.898 g/mL at 25 °C(lit.)

Description

Caution

may contain precipitate

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
80.6 °F
Flash Point(C) 
27 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?