EMAIL THIS PAGE TO A FRIEND
93430 Sigma-Aldrich

Tritriacontane

purum, ≥97.0% (GC)

  • CAS Number 630-05-7

  • Linear Formula CH3(CH2)31CH3

  • Molecular Weight 464.89

  •  Beilstein Registry Number 1786920

  •  MDL number MFCD00010187

  •  PubChem Substance ID 329770233

Purchase

Properties

Related Categories Acyclic, Alkanes, Building Blocks, Cell Biology, Chemical Synthesis,
InChI Key   SUJUOAZFECLBOA-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
mp   71-72 °C
  71-73 °C(lit.)

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?