• USA Home
  • 93463 - (S)-(+)-γ-Trityloxymethyl-γ-butyrolactone

EMAIL THIS PAGE TO A FRIEND
93463 Sigma-Aldrich

(S)-(+)-γ-Trityloxymethyl-γ-butyrolactone

purum, ≥99.0% (CH)

Synonym: (S)-(+)-Dihydro-5-trityloxymethyl-2(3H)-furanone, 5-O-Trityl-2,3-dideoxy-D-glyceropentono-1,4-lactone

  • CAS Number 73968-62-4

  • Empirical Formula (Hill Notation) C24H22O3

  • Molecular Weight 358.43

  •  Beilstein Registry Number 4715758

  •  MDL number MFCD00066260

  •  PubChem Substance ID 329770238

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Lactones, Organic Building Blocks More...
InChI Key   OZWWLABCDGFABG-QFIPXVFZSA-N
grade   purum
assay   ≥99.0% (CH)
optical activity   [α]/D +27.0±1.0°, c = 1 in methylene chloride
mp   150-152 °C(lit.)
  150-152 °C

Description

Other Notes

Chiral building block in the synthesis of various compounds

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?