• USA Home
  • 93464 - Tritylpyridinium tetrafluoroborate

EMAIL THIS PAGE TO A FRIEND
93464 Sigma-Aldrich

Tritylpyridinium tetrafluoroborate

purum

Synonym: 1-(Triphenylmethyl)pyridinium tetrafluoroborate

  • CAS Number 26156-84-3

  • Empirical Formula (Hill Notation) C24H20BF4N

  • Molecular Weight 409.23

  •  Beilstein Registry Number 3834611

  •  EC Number 247-482-0

  •  MDL number MFCD00042587

  •  PubChem Substance ID 329770239

Purchase

Properties

Related Categories Building Blocks, C16+, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   PBUNNYDYPYZWML-UHFFFAOYSA-N
grade   purum
mp   175-181 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Potent tritylating agent with several advantages over the classical trityl chloride (more reactive, high selectivity, simple manipulations)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?