EMAIL THIS PAGE TO A FRIEND
93680 Sigma-Aldrich

DL-Tryptophan

puriss., ≥99.0% (NT)

Synonym: (±)-α-Amino-3-indolepropionic acid, (±)-2-Amino-3-(3-indolyl)propionic acid, DL-3β-Indolylalanine

  • CAS Number 54-12-6

  • Empirical Formula (Hill Notation) C11H12N2O2

  • Molecular Weight 204.23

  •  Beilstein Registry Number 86199

  •  EC Number 200-194-9

  •  MDL number MFCD00064339

  •  PubChem Substance ID 329770258

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Tryptophan More...
InChI Key   QIVBCDIJIAJPQS-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
mp   289-290 °C (dec.)(lit.)
  ~295 °C (dec.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Tryptophan Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Tryptophan Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?