• USA Home
  • 93755 - Tunicamycin from Streptomyces sp.

EMAIL THIS PAGE TO A FRIEND
93755 Sigma-Aldrich

Tunicamycin from Streptomyces sp.

Purchase

Properties

Related Categories Antibacterial, Antibiotics, Antibiotics A to Z, Antibiotics T-Z, Antibiotics by Application,
InChI Key   YJQCOFNZVFGCAF-WPTOCQRYSA-N
Mode of action   protein synthesis | interferes
antibiotic activity spectrum   fungi
  viruses
storage temp.   2-8°C

Description

Other Notes

Glucosamine containing antibiotic with cell-surface altering properties, enhances the antiviral and antigrowth effects of interferon; Many important biochemical functions are inhibited by tunicamycin, such as glycoprotein biosynthesis in yeast; The synthesis of polyisoprenol sugars in bacteria

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

General description

Mixture of homologous tunicamycins, which differ in the aliphatic side chain

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3462 6.1 / PGII
WGK Germany 
3
RTECS 
YO7980200

Documents

Certificate of Analysis

Protocols & Articles

Articles

Inhibition of Cell Wall Biosynthesis by Antibiotics

Antibiotics are molecules that specifically target and kill cells. The term antibiotic is frequently used interchangeably with the word antibacterial, but antiviral, antifungal and antineoplastic com...
BioFiles 2006, 1.4, 7.
Keywords: Antibiotics, Antifungals, Antivirals, Building blocks, Glycosylations, Metabolism

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?