EMAIL THIS PAGE TO A FRIEND
93840 Sigma-Aldrich

D-Tyrosine

puriss., ≥99.0% (NT)

Synonym: (R)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-D-alanine

  • CAS Number 556-02-5

  • Linear Formula 4-(HO)C6H4CH2CH(NH2)CO2H

  • Molecular Weight 181.19

  •  Beilstein Registry Number 2212157

  •  EC Number 209-112-6

  •  MDL number MFCD00063073

  •  PubChem Substance ID 329770277

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Tyrosine More...
InChI Key   OUYCCCASQSFEME-MRVPVSSYSA-N
grade   puriss.
assay   ≥99.0% (NT)
optical activity   [α]20/D +11.5±1.0°, c = 4% in 1 M HCl
mp   >300 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

HPLC Analysis of Tyrosine Enantiomers on Astec® CHIROBIOTIC® T

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Tyrosine Enantiomers on Astec® CHIROBIOTIC® T
Keywords: Chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?