EMAIL THIS PAGE TO A FRIEND
93850 Sigma-Aldrich

DL-Tyrosine

puriss., ≥99.0% (NT)

Synonym: (±)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-DL-alanine

  • CAS Number 556-03-6

  • Linear Formula 4-(HO)C6H4CH2CH(NH2)COOH

  • Molecular Weight 181.19

  •  Beilstein Registry Number 515881

  •  EC Number 209-113-1

  •  MDL number MFCD00063074

  •  PubChem Substance ID 24889934

Purchase

Properties

Related Categories Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry, Tyrosine More...
InChI Key   OUYCCCASQSFEME-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
mp   ≥300 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

145726 DL-Tyrosine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?