EMAIL THIS PAGE TO A FRIEND
94040 Sigma-Aldrich

Undecanedioic acid

puriss., ≥99.0% (GC)

Synonym: 1,9-Nonanedicarboxylic acid

Purchase

Properties

Related Categories Building Blocks, C11 to C12, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   LWBHHRRTOZQPDM-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
mp   108-110 °C(lit.)
  110-111 °C
solubility   methanol: soluble10 mg/mL, clear

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

177962 Undecanedioic acid, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?