EMAIL THIS PAGE TO A FRIEND
94061 Sigma-Aldrich

1-Undecanol

purum, ≥98.0% (GC)

Synonym: Alcohol C11, Undecyl alcohol

  • CAS Number 112-42-5

  • Linear Formula CH3(CH2)10OH

  • Molecular Weight 172.31

  •  Beilstein Registry Number 1698334

  •  EC Number 203-970-5

  •  MDL number MFCD00004751

  •  PubChem Substance ID 329770293

Purchase

Properties

Related Categories Alcohols, Building Blocks, C11 to C30+, Chemical Synthesis, Organic Building Blocks,
InChI Key   KJIOQYGWTQBHNH-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.44(lit.)
  n20/D 1.441
bp   146 °C/30 mmHg(lit.)
mp   11 °C(lit.)
density   0.83 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
3
RTECS 
YQ3155000
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

U1001 1-Undecanol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?