• USA Home
  • 94205 - C-Undecylcalix[4]resorcinarene monohydrate

EMAIL THIS PAGE TO A FRIEND
94205 Sigma-Aldrich

C-Undecylcalix[4]resorcinarene monohydrate

puriss., ≥99.0% (HPLC)

  • CAS Number 112247-07-1

  • Empirical Formula (Hill Notation) C72H112O8 · H2O

  • Molecular Weight 1123.67

  •  Beilstein Registry Number 4290394

  •  MDL number MFCD00192571

  •  PubChem Substance ID 329770312

Purchase

Properties

Related Categories Calixarenes, Chelation/Complexation Compounds, Chemical Synthesis, Synthetic Reagents
InChI Key   JKLZYDGPENNXSJ-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (HPLC)
mp   293-295 °C(lit.)
  293-295 °C

Description

Other Notes

Lipophilic polar host with four independent binding sites. Carboxylic acids, sugars and other polyhydroxy compounds are complexed very selectively; Stereochemical assignment of chiral guests

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

414824 C-Undecylcalix[4]resorcinarene monohydrate, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?