EMAIL THIS PAGE TO A FRIEND
94220 Sigma-Aldrich

Uracil

≥99.0% (T)

Synonym: 2,4(1H,3H)-Pyrimidinedione, 2,4-Dihydroxypyrimidine, 2,4-Pyrimidinediol

  • CAS Number 66-22-8

  • Empirical Formula (Hill Notation) C4H4N2O2

  • Molecular Weight 112.09

  •  Beilstein Registry Number 507828

  •  EC Number 200-621-9

  •  MDL number MFCD00006016

  •  PubChem Substance ID 329770314

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Cell Biology, Chemical Synthesis, Heterocyclic Building Blocks,
InChI Key   ISAKRJDGNUQOIC-UHFFFAOYSA-N
assay   ≥99.0% (T)
ign. residue   ≤0.1%
mp   >300 °C(lit.)

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
YQ8650000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

U0750 Uracil, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?