• USA Home
  • 94531 - Dowex® Monosphere® 650C

EMAIL THIS PAGE TO A FRIEND
94531 Sigma-Aldrich

Dowex® Monosphere® 650C

hydrogen form, 25-30 mesh

Purchase

Properties

Related Categories Analytical/Chromatography, Cation, LPLC Separation Media, LPLC/TLC/Paper Chromatography, Miscellaneous More...
parameter   150 °C max. temp.
cross-linkage   8%
moisture   46-51%
loss   ~50% loss on drying, 110 °C
matrix   styrene-divinylbenzene (gel)
particle size   25-30 mesh
operating pH   0 - 14
capacity   2.0 meq/mL by wetted bed volume
bulk density   690 g/L

Description

Other Notes

replaces Cat. No. 44500

Application

Strongly acidic cation exchange resin for mixed-bed applications. Commonly used for condensate polishing and sugar applications. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Legal Information

DOWEX is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Monosphere is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?