EMAIL THIS PAGE TO A FRIEND
94674 Sigma-Aldrich

D-Valinol

purum, ≥96.0% (sum of enantiomers, GC)

Synonym: (R)-2-Amino-3-methyl-1-butanol

  • CAS Number 4276-09-9

  • Empirical Formula (Hill Notation) C5H13NO

  • Molecular Weight 103.16

  •  Beilstein Registry Number 1719138

  •  MDL number MFCD00064297

  •  PubChem Substance ID 329770373

Purchase

Properties

Related Categories Amino Acids & Derivatives, Amino Alcohols, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   NWYYWIJOWOLJNR-YFKPBYRVSA-N
grade   purum
assay   ≥96.0% (sum of enantiomers, GC)
optical activity   [α]20/D −10±1°, c = 10% in H2O
refractive index   n20/D 1.455
bp   189-190 °C(lit.)
mp   30-34 °C
density   0.931 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
172.4 °F
Flash Point(C) 
78 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

284483 (R)-(−)-2-Amino-3-methyl-1-butanol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?