• USA Home
  • 94680 - D-Val-Leu-Lys 4-nitroanilide dihydrochloride

EMAIL THIS PAGE TO A FRIEND
94680 Sigma-Aldrich

D-Val-Leu-Lys 4-nitroanilide dihydrochloride

≥98.0% (HPLC)

Synonym: D-Val-Leu-Lys-pNA dihydrochloride, D-Valyl-L-leucyl-L-lysine 4-nitroanilide dihydrochloride

  • CAS Number 62354-43-2

  • Empirical Formula (Hill Notation) C23H38N6O5 · 2HCl

  • Molecular Weight 551.51

  •  MDL number MFCD00077899

  •  PubChem Substance ID 329770377

Purchase

Properties

Related Categories Biochemicals and Reagents, Enzyme Substrates, Enzymes, Inhibitors, and Substrates, Plasmin, Substrates by Enzyme More...
InChI Key   VESQMNNSPPEOSZ-ZLARAOTRSA-N
assay   ≥98.0% (HPLC)
suitability   suitable as substrate for plasmin test
storage temp.   −20°C

Description

Other Notes

Substrate for plasmin; Preparation and purification of microplasmin

Price and Availability


Biomedical Applications
Safety & Documentation

Safety Information

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

V0882 D-Val-Leu-Lys 4-nitroanilide dihydrochloride, plasmin substrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?