EMAIL THIS PAGE TO A FRIEND
94720 Sigma-Aldrich

Vanadium(V) oxide

purum, ≥98.0% (RT)

Synonym: Divanadium pentaoxide, Divanadium pentoxide, Pentaoxodivanadium, Vandia

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Vanadium More...
InChI Key   GNTDGMZSJNCJKK-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (RT)
mp   690 °C(lit.)
density   3.35 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 2862 6.1 / PGIII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

223794 Vanadium(V) oxide, ≥98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?