EMAIL THIS PAGE TO A FRIEND
94840 Sigma-Aldrich

1,2-Dimethoxybenzene

puriss., ≥99.0% (GC)

Synonym: Pyrocatechol dimethyl ether, Veratrole

  • CAS Number 91-16-7

  • Linear Formula C6H4(OCH3)2

  • Molecular Weight 138.16

  •  Beilstein Registry Number 1364621

  •  EC Number 202-045-3

  •  MDL number MFCD00008357

  •  PubChem Substance ID 24890065

Purchase

Properties

Related Categories Building Blocks, C8, Chemical Synthesis, Ethers, Organic Building Blocks,
InChI Key   ABDKAPXRBAPSQN-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (GC)
refractive index   n20/D 1.533(lit.)
  n20/D 1.534
bp   206-207 °C(lit.)
mp   15 °C(lit.)
  22-23 °C
density   1.084 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
WGK Germany 
1
RTECS 
CZ6475000
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

140155 1,2-Dimethoxybenzene, ReagentPlus®, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?