EMAIL THIS PAGE TO A FRIEND
94965 Sigma-Aldrich

Vinylene carbonate

purum, ≥97.0% (GC)

Synonym: 1,3-Dioxol-2-one, VC

  • CAS Number 872-36-6

  • Empirical Formula (Hill Notation) C3H2O3

  • Molecular Weight 86.05

  •  Beilstein Registry Number 105683

  •  MDL number MFCD00005380

  •  PubChem Substance ID 24889967

Purchase

Properties

Related Categories Building Blocks, Carbonates, Carbonyl Compounds, Chemical Synthesis, Organic Building Blocks More...
InChI Key   VAYTZRYEBVHVLE-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
contains   ~0.5% 2,6-di-tert-butyl-4-methylphenol as stabilizer
refractive index   n20/D 1.421(lit.)
  n20/D 1.422
bp   162 °C(lit.)
mp   19-22 °C(lit.)
density   1.360 g/mL at 20 °C
  1.355 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
RIDADR 
UN2810 - class 6.1 - PG 3 - EHS - Toxic, liquids, organic, n.o.s., HI: all
WGK Germany 
3
RTECS 
FG3325000
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

V2607 Vinylene carbonate, contains ≤2% BHT as stabilizer, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?