• USA Home
  • 94975 - Vinylene trithiocarbonate

EMAIL THIS PAGE TO A FRIEND
94975 Sigma-Aldrich

Vinylene trithiocarbonate

purum, ≥99.0% (GC), yellow

Synonym: 1,3-Dithiole-2-thione

  • CAS Number 930-35-8

  • Empirical Formula (Hill Notation) C3H2S3

  • Molecular Weight 134.24

  •  Beilstein Registry Number 105686

  •  EC Number 213-215-1

  •  MDL number MFCD00014536

  •  PubChem Substance ID 329770407

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiocarbonyl Compounds More...
InChI Key   WYKJWNVWJOKVQP-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
color   yellow
mp   48-50 °C(lit.)
  48-50 °C

Description

Other Notes

Key intermediate in TTF synthesis

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
JP1292125
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

358916 Vinylene trithiocarbonate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?