Skip to Content
MilliporeSigma

95060

1-Vinyl-2-pyrrolidinone

purum, ≥97.0% (GC)

Synonym(s):

1-Vinyl-2-pyrrolidone

Sign In to View Organizational & Contract Pricing

About This Item

Empirical Formula (Hill Notation):
C6H9NO
CAS Number:
Molecular Weight:
111.14
Beilstein/REAXYS Number:
110513
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor density

3.8 (vs air)

vapor pressure

0.1 mmHg ( 24 °C)

grade

purum

assay

≥97.0% (GC)

autoignition temp.

685 °F

contains

~0.001% N,N′-di-sec-butyl-p-phenylenediamine as stabilizer

expl. lim.

10 %

refractive index

n20/D 1.512 (lit.)
n20/D 1.513

bp

92-95 °C/11 mmHg (lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

[O-][N+](=O)\C=C\c1ccc(F)cc1
C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

comparable product

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - STOT RE 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

203.0 °F - closed cup

flash_point_c

95 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana C Borges et al.
Acta biomaterialia, 7(9), 3412-3421 (2011-06-10)
The swelling and compressive mechanical behavior as well as the morphology and biocompatibility of composite hydrogels based on Tween® 20 trimethacrylate (T3), N-vinyl-2-pyrrolidone (NVP) and nanofibrillated cellulose (NFC) were assessed in the present study. The chemical structure of T3 was
Zhaoqiang Wu et al.
Colloids and surfaces. B, Biointerfaces, 96, 37-43 (2012-04-19)
A new method for the modification of poly(dimethylsiloxane) (PDMS) elastomer surfaces with hydrophilic poly(N-vinylpyrrolidone) (PVP) has been developed. PVP chains were grafted from the PDMS surface by surface-initiated atom transfer radical polymerization (SI-ATRP). The resulting surfaces were characterized by X-ray
Volker Engels et al.
Dalton transactions (Cambridge, England : 2003), 39(28), 6496-6502 (2010-06-23)
A modified polyol-based reduction method in ethylene glycol that incorporates poly(N-vinylpyrrolidone) (PVP, M(av) = 10,000; 40,000; 55,000) as polymeric anti-agglomerant alongside a reducing additive (N(2)H(4) x H(2)O, NaBH(4), NaH(2)PO(2) x H(2)O) has been employed to investigate the influence of synthetic
Hao Jiang et al.
Journal of chromatography. A, 1246, 111-116 (2012-03-27)
In this work, a novel kind of N-vinyl-2-pyrrolidinone (NVP) modified poly acrylic ester microspheres was prepared, followed by trypsin immobilization to prepare a hydrophilic immobilized enzyme reactor (IMER), to achieve highly efficient protein digestion with low peptide residue. The nonspecific
Nadège Handké et al.
Colloids and surfaces. B, Biointerfaces, 103, 298-303 (2013-01-02)
Efficient biomolecule conjugation to the surface of biodegradable colloidal carriers is crucial for their targeting efficiency in drug/vaccine delivery applications. We here propose a potent strategy to drastically improve peptide immobilization on biodegradable polylactide (PLA) nanoparticles (NPs). Our approach particularly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service