• USA Home
  • 95085 - Tributyl(vinyl)stannane

EMAIL THIS PAGE TO A FRIEND
95085 Sigma-Aldrich

Tributyl(vinyl)stannane

purum, ≥97.0% (GC)

Synonym: Tributylstannylethylene, Vinyltributylstannane

  • CAS Number 7486-35-3

  • Linear Formula CH2=CHSn[CH3(CH2)3]3

  • Molecular Weight 317.10

  •  Beilstein Registry Number 3537662

  •  EC Number 231-291-4

  •  MDL number MFCD00009421

  •  PubChem Substance ID 329770417

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organotin, Organotins
InChI Key   QIWRFOJWQSSRJZ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.478(lit.)
bp   104-106 °C/3.5 mmHg(lit.)
density   1.085 g/mL at 25 °C(lit.)

Description

Other Notes

Reagent used for the Pd-catalyzed Stille cross-coupling vinylation ;

Same product — same quality — now Sigma-Aldrich brand

Price and Availability


Cross-Coupling Guide
Safety & Documentation

Safety Information

Signal word 
Danger
RIDADR 
UN 2788 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
109.4 °F
Flash Point(C) 
43 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?