• USA Home
  • 95100 - Trimethyl(vinyl)silane

EMAIL THIS PAGE TO A FRIEND
95100 Sigma-Aldrich

Trimethyl(vinyl)silane

purum, ≥97.0% (GC)

Synonym: (Trimethylsilyl)ethylene, Ethenyltrimethylsilane

  • CAS Number 754-05-2

  • Linear Formula (CH3)3SiCHCH2

  • Molecular Weight 100.23

  •  Beilstein Registry Number 956572

  •  EC Number 212-042-9

  •  MDL number MFCD00008606

Purchase

Properties

Related Categories Chemical Synthesis, Organometallic Reagents, Organosilicon, Other Organosilicon
vapor pressure   4.44 psi ( 20 °C)
InChI Key   GCSJLQSCSDMKTP-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.391(lit.)
  n20/D 1.391
bp   55 °C(lit.)
density   0.684 g/mL at 25 °C(lit.)

Description

Other Notes

Review

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
3
Flash Point(F) 
-11.2 °F
Flash Point(C) 
-24 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?